Product Details
- SNP ID
-
rs1298094
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72838251 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCAGGGCAGTCCAATTTTTCCC[A/C]ATAAGGTTTCACTTTCTCAGTCTGG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATG16L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76762469] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATG16L2
- Gene Name
- autophagy related 16 like 2
- Gene
- FCHSD2
- Gene Name
- FCH and double SH3 domains 2
View Full Product Details