Product Details

SNP ID
rs951833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116645794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTTAAATTTCCCATAGATTTCC[A/G]AAGTTGCTTGAGACTGCTAGCCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZUFSP PubMed Links

Gene Details

Gene
ZUFSP
Gene Name
zinc finger with UFM1 specific peptidase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145062.2 2262 Missense Mutation CGG,TGG R537W NP_659499.2
XM_011535565.2 2262 Missense Mutation CGG,TGG R537W XP_011533867.1
XM_011535567.1 2262 Missense Mutation CGG,TGG R369W XP_011533869.1
XM_017010393.1 2262 Missense Mutation CGG,TGG R537W XP_016865882.1
XM_017010394.1 2262 Missense Mutation CGG,TGG R341W XP_016865883.1
XM_017010395.1 2262 Missense Mutation CGG,TGG R313W XP_016865884.1
XM_017010396.1 2262 Missense Mutation CGG,TGG R313W XP_016865885.1

View Full Product Details