Product Details

SNP ID
rs807181
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:108090354 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCATGTTTTTTACGTCTCTTGAT[C/G]CTGAATAGTTCCTTTCCCTTTTTAA
Phenotype
MIM: 300663 MIM: 300880
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ATG4A PubMed Links

Gene Details

Gene
ATG4A
Gene Name
autophagy related 4A cysteine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321287.1 Intron NP_001308216.1
NM_001321288.1 Intron NP_001308217.1
NM_001321289.1 Intron NP_001308218.1
NM_001321290.1 Intron NP_001308219.1
NM_052936.4 Intron NP_443168.2
NM_178270.3 Intron NP_840054.1
NM_178271.2 Intron NP_840055.1
XM_011530841.2 Intron XP_011529143.1
XM_011530842.1 Intron XP_011529144.1
Gene
PSMD10
Gene Name
proteasome 26S subunit, non-ATPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002814.3 Intron NP_002805.1
NM_170750.2 Intron NP_736606.1

View Full Product Details