Product Details

SNP ID
rs974722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1801822 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTTGCTGTGGACTGGCTGCCCAT[A/G]GGCGATTTGTGGCAGCCGATTATTT
Phenotype
MIM: 611842 MIM: 603848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL36 PubMed Links

Gene Details

Gene
MRPL36
Gene Name
mitochondrial ribosomal protein L36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032479.3 Intron NP_115868.1
XM_011514079.2 Intron XP_011512381.1
XM_011514080.2 Intron XP_011512382.1
XM_017009750.1 Intron XP_016865239.1
XM_017009751.1 Intron XP_016865240.1
XM_017009752.1 Intron XP_016865241.1
Gene
NDUFS6
Gene Name
NADH:ubiquinone oxidoreductase subunit S6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004553.4 Intron NP_004544.1

View Full Product Details