Product Details
- SNP ID
-
rs1046425
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141515086 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAACTGGAGGTGATGACATCCCA[A/G]AAGCCTATGAGGCAAGGGAAATAGG
- Phenotype
-
MIM: 602121
MIM: 606288
MIM: 606297
MIM: 606298
MIM: 603059
MIM: 606289
MIM: 606290
MIM: 606291
MIM: 606292
MIM: 606293
MIM: 606294
MIM: 606295
MIM: 606296
MIM: 606299
MIM: 606300
MIM: 606301
MIM: 603058
MIM: 606302
MIM: 606303
MIM: 606304
MIM: 603627
MIM: 606305
MIM: 606306
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DIAPH1
PubMed Links
Gene Details
- Gene
- DIAPH1
- Gene Name
- diaphanous related formin 1
- Gene
- PCDHGA1
- Gene Name
- protocadherin gamma subfamily A, 1
There are no transcripts associated with this gene.
- Gene
- PCDHGA10
- Gene Name
- protocadherin gamma subfamily A, 10
There are no transcripts associated with this gene.
- Gene
- PCDHGA11
- Gene Name
- protocadherin gamma subfamily A, 11
There are no transcripts associated with this gene.
- Gene
- PCDHGA12
- Gene Name
- protocadherin gamma subfamily A, 12
There are no transcripts associated with this gene.
- Gene
- PCDHGA2
- Gene Name
- protocadherin gamma subfamily A, 2
There are no transcripts associated with this gene.
- Gene
- PCDHGA3
- Gene Name
- protocadherin gamma subfamily A, 3
There are no transcripts associated with this gene.
- Gene
- PCDHGA4
- Gene Name
- protocadherin gamma subfamily A, 4
There are no transcripts associated with this gene.
- Gene
- PCDHGA5
- Gene Name
- protocadherin gamma subfamily A, 5
There are no transcripts associated with this gene.
- Gene
- PCDHGA6
- Gene Name
- protocadherin gamma subfamily A, 6
There are no transcripts associated with this gene.
- Gene
- PCDHGA7
- Gene Name
- protocadherin gamma subfamily A, 7
There are no transcripts associated with this gene.
- Gene
- PCDHGA8
- Gene Name
- protocadherin gamma subfamily A, 8
There are no transcripts associated with this gene.
- Gene
- PCDHGA9
- Gene Name
- protocadherin gamma subfamily A, 9
There are no transcripts associated with this gene.
- Gene
- PCDHGB1
- Gene Name
- protocadherin gamma subfamily B, 1
There are no transcripts associated with this gene.
- Gene
- PCDHGB2
- Gene Name
- protocadherin gamma subfamily B, 2
There are no transcripts associated with this gene.
- Gene
- PCDHGB3
- Gene Name
- protocadherin gamma subfamily B, 3
There are no transcripts associated with this gene.
- Gene
- PCDHGB4
- Gene Name
- protocadherin gamma subfamily B, 4
There are no transcripts associated with this gene.
- Gene
- PCDHGB5
- Gene Name
- protocadherin gamma subfamily B, 5
There are no transcripts associated with this gene.
- Gene
- PCDHGB6
- Gene Name
- protocadherin gamma subfamily B, 6
There are no transcripts associated with this gene.
- Gene
- PCDHGB7
- Gene Name
- protocadherin gamma subfamily B, 7
There are no transcripts associated with this gene.
- Gene
- PCDHGC3
- Gene Name
- protocadherin gamma subfamily C, 3
There are no transcripts associated with this gene.
- Gene
- PCDHGC4
- Gene Name
- protocadherin gamma subfamily C, 4
There are no transcripts associated with this gene.
- Gene
- PCDHGC5
- Gene Name
- protocadherin gamma subfamily C, 5
There are no transcripts associated with this gene.
View Full Product Details