Product Details

SNP ID
rs1042647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42161234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGAGTGTGATGTGAGAACTTAA[C/G]GAAGTGCTGGCATGGGCAGGCACGC
Phenotype
MIM: 603107
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCF20 PubMed Links

Gene Details

Gene
TCF20
Gene Name
transcription factor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005650.3 6373 UTR 3 NP_005641.1
NM_181492.2 6373 UTR 3 NP_852469.1
XM_005261722.3 6373 UTR 3 XP_005261779.1
XM_006724313.3 6373 UTR 3 XP_006724376.1
XM_011530353.2 6373 UTR 3 XP_011528655.1
XM_011530354.2 6373 UTR 3 XP_011528656.1

View Full Product Details