Product Details

SNP ID
rs6684320
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:62204237 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGTTTTTCATTTCTTTGATTTG[C/T]GCGCATCTCTTTTGGTTTCTTGGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
L1TD1 PubMed Links

Gene Details

Gene
L1TD1
Gene Name
LINE1 type transposase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details