Product Details

SNP ID
rs11855621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65124846 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAAAGCAAAACCCCAACAGCCCC[A/G]TGCTCTGTGCTCACCTGTCAGCTAC
Phenotype
MIM: 608138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDCD7 PubMed Links

Gene Details

Gene
PDCD7
Gene Name
programmed cell death 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005707.1 Intron NP_005698.1

View Full Product Details