Product Details

SNP ID
rs34057707
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:81317360 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCTGAAAATTCCTCTCTGTCAGACC[C/T]GGCACCACTGAAAGGTGAGTCTTAG
Phenotype
MIM: 603035 MIM: 607050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL16 PubMed Links

Gene Details

Gene
IL16
Gene Name
interleukin 16
There are no transcripts associated with this gene.

Gene
STARD5
Gene Name
StAR related lipid transfer domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181900.2 Intron NP_871629.1
XM_006720703.1 Intron XP_006720766.1
XM_017022639.1 Intron XP_016878128.1
Gene
TMC3-AS1
Gene Name
TMC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details