Product Details

SNP ID
rs1342785
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:90713093 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCAGGCTGTGAGAGAAGAAACC[A/G]TAAACAAAGGGATGCAGAGACTCCT
Phenotype
MIM: 605212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BARHL2 PubMed Links

Gene Details

Gene
BARHL2
Gene Name
BarH like homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020063.1 Intron NP_064447.1

View Full Product Details