Product Details

SNP ID
rs903209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:12286953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGTATGGTGTGAGGGAAGGAGAG[A/G]GTTCAGTCTATTATAATCATTAGTC
Phenotype
MIM: 608881 MIM: 612355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICAL2 PubMed Links

Gene Details

Gene
MICAL2
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 2
There are no transcripts associated with this gene.

Gene
MICALCL
Gene Name
MICAL C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032867.2 54 UTR 5 NP_116256.2

View Full Product Details