Product Details

SNP ID
rs1389550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:12291024 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGTGGATGAAAATCTGAAGGCTA[C/G]ACACCCTGCAGGAAAATCTGAATGG
Phenotype
MIM: 612355
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MICALCL PubMed Links

Gene Details

Gene
MICALCL
Gene Name
MICAL C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032867.2 Intron NP_116256.2

View Full Product Details