Product Details

SNP ID
rs1162763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68333280 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATAATTGTTGCTTTTTGAACTTT[C/G]GGGGTGGGGGTGGGTGATGTAGCAA
Phenotype
MIM: 602324 MIM: 612189 MIM: 610328
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPH3 PubMed Links

Gene Details

Gene
HNRNPH3
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322434.1 Intron NP_001309363.1
NM_001322436.1 Intron NP_001309365.1
NM_001322437.1 Intron NP_001309366.1
NM_001322438.1 Intron NP_001309367.1
NM_001322439.1 Intron NP_001309368.1
NM_001322440.1 Intron NP_001309369.1
NM_001322441.1 Intron NP_001309370.1
NM_001322442.1 Intron NP_001309371.1
NM_001322443.1 Intron NP_001309372.1
NM_001322444.1 Intron NP_001309373.1
NM_001322445.1 Intron NP_001309374.1
NM_001322446.1 Intron NP_001309375.1
NM_001322447.1 Intron NP_001309376.1
NM_001322448.1 Intron NP_001309377.1
NM_001322449.1 Intron NP_001309378.1
NM_001322450.1 Intron NP_001309379.1
NM_001322451.1 Intron NP_001309380.1
NM_001322452.1 Intron NP_001309381.1
NM_001322453.1 Intron NP_001309382.1
NM_012207.2 Intron NP_036339.1
NM_021644.3 Intron NP_067676.2
XM_011539742.1 Intron XP_011538044.1
XM_017016168.1 Intron XP_016871657.1
XM_017016169.1 Intron XP_016871658.1
Gene
PBLD
Gene Name
phenazine biosynthesis like protein domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033083.1 Intron NP_001028255.1
NM_022129.3 Intron NP_071412.2
XM_005270028.3 Intron XP_005270085.1
XM_011540060.2 Intron XP_011538362.1
XM_017016513.1 Intron XP_016872002.1
XM_017016514.1 Intron XP_016872003.1
Gene
RUFY2
Gene Name
RUN and FYVE domain containing 2
There are no transcripts associated with this gene.

View Full Product Details