Product Details

SNP ID
rs12285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71859824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCCCTTGTCCAGAGCTGCCTT[G/T]AGCCAGGTCCCTCCTGACCCCATCA
Phenotype
MIM: 614301
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATXN1L PubMed Links

Gene Details

Gene
ATXN1L
Gene Name
ataxin 1 like
There are no transcripts associated with this gene.

Gene
ZNF821
Gene Name
zinc finger protein 821
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201552.1 1371 UTR 3 NP_001188481.1
NM_001201553.1 1371 UTR 3 NP_001188482.1
NM_001201554.1 1371 UTR 3 NP_001188483.1
NM_001201556.1 1371 UTR 3 NP_001188485.1
NM_001318238.1 1371 UTR 3 NP_001305167.1
NM_001318239.1 1371 UTR 3 NP_001305168.1
NM_017530.2 1371 UTR 3 NP_060000.1
XM_005256033.2 1371 UTR 3 XP_005256090.1
XM_006721233.1 1371 UTR 3 XP_006721296.1
XM_011523211.2 1371 UTR 3 XP_011521513.1
XM_011523212.2 1371 UTR 3 XP_011521514.1
XM_011523214.2 1371 UTR 3 XP_011521516.1
XM_011523215.2 1371 UTR 3 XP_011521517.1
XM_011523220.2 1371 UTR 3 XP_011521522.1
XM_017023409.1 1371 UTR 3 XP_016878898.1
XM_017023410.1 1371 UTR 3 XP_016878899.1
XM_017023411.1 1371 UTR 3 XP_016878900.1
XM_017023412.1 1371 UTR 3 XP_016878901.1
XM_017023413.1 1371 UTR 3 XP_016878902.1
XM_017023414.1 1371 UTR 3 XP_016878903.1
XM_017023415.1 1371 UTR 3 XP_016878904.1
XM_017023416.1 1371 UTR 3 XP_016878905.1
XM_017023417.1 1371 UTR 3 XP_016878906.1
XM_017023418.1 1371 UTR 3 XP_016878907.1
XM_017023419.1 1371 UTR 3 XP_016878908.1
XM_017023420.1 1371 UTR 3 XP_016878909.1
XM_017023421.1 1371 UTR 3 XP_016878910.1
XM_017023422.1 1371 UTR 3 XP_016878911.1
XM_017023423.1 1371 UTR 3 XP_016878912.1
XM_017023424.1 1371 UTR 3 XP_016878913.1
XM_017023425.1 1371 UTR 3 XP_016878914.1

View Full Product Details