Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012614.1 | 206 | Intron | NP_001012632.1 | ||
NM_001328.2 | 206 | Intron | NP_001319.1 | ||
XM_005272261.3 | 206 | Intron | XP_005272318.1 | ||
XM_005272263.4 | 206 | Intron | XP_005272320.1 | ||
XM_011513395.2 | 206 | Intron | XP_011511697.1 | ||
XM_011513396.2 | 206 | Intron | XP_011511698.1 | ||
XM_011513397.2 | 206 | Intron | XP_011511699.1 | ||
XM_017007762.1 | 206 | Intron | XP_016863251.1 | ||
XM_017007763.1 | 206 | UTR 5 | XP_016863252.1 | ||
XM_017007764.1 | 206 | Intron | XP_016863253.1 | ||
XM_017007765.1 | 206 | Intron | XP_016863254.1 | ||
XM_017007766.1 | 206 | Intron | XP_016863255.1 | ||
XM_017007767.1 | 206 | Intron | XP_016863256.1 |