Product Details

SNP ID
rs930808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73395182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAATGGGAGTGGCTGGCAGGGC[A/G]GGATAGGTGGGGCCAGGGGCGGAGC
Phenotype
MIM: 606286
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX43 PubMed Links

Gene Details

Gene
DDX43
Gene Name
DEAD-box helicase 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018665.2 Intron NP_061135.2
XM_011535926.1 Intron XP_011534228.1
XM_011535927.2 Intron XP_011534229.1
XM_011535928.2 Intron XP_011534230.1
XM_017011003.1 Intron XP_016866492.1
Gene
OOEP-AS1
Gene Name
OOEP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details