Product Details

SNP ID
rs6792482
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184036241 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTACAAACCCCCAGAATATGAT[C/T]ATAGGTAGAAGAGAGCAGTCATCTG
Phenotype
MIM: 610122
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HTR3D PubMed Links

Gene Details

Gene
HTR3D
Gene Name
5-hydroxytryptamine receptor 3D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145143.1 Intron NP_001138615.1
NM_001163646.1 Intron NP_001157118.1
NM_182537.2 Intron NP_872343.2
XM_017005854.1 Intron XP_016861343.1

View Full Product Details