Product Details

SNP ID
rs35282341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158947083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTTCCTTCCAATCTACATGGA[C/T]TCTCATCCATTCTTCTAACTGAGCA
Phenotype
MIM: 615876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSPH3 PubMed Links

Gene Details

Gene
RSPH3
Gene Name
radial spoke 3 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031924.4 Intron NP_114130.3
XM_005267153.4 Intron XP_005267210.1
XM_017011347.1 Intron XP_016866836.1

View Full Product Details