Product Details

SNP ID
rs5930434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:131074245 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACAGATGTGGTCTGGAGAGGCA[A/G]ATATTGATGACCCTTCTTTTAATTA
Phenotype
MIM: 300937
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP36 PubMed Links

Gene Details

Gene
ARHGAP36
Gene Name
Rho GTPase activating protein 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282607.1 Intron NP_001269536.1
NM_144967.3 Intron NP_659404.2
XM_006724729.1 Intron XP_006724792.1
XM_011531280.1 Intron XP_011529582.1

View Full Product Details