Product Details

SNP ID
rs3124591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136495945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATACTAAAAAAAATTAAAATCCT[C/T]GTTCTTATTTTGTATAAAAACATGT
Phenotype
MIM: 190198
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf163 PubMed Links

Gene Details

Gene
C9orf163
Gene Name
chromosome 9 open reading frame 163
There are no transcripts associated with this gene.

Gene
NOTCH1
Gene Name
notch 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017617.4 7086 UTR 3 NP_060087.3
XM_011518717.2 7086 UTR 3 XP_011517019.2

View Full Product Details