Product Details

SNP ID
rs3744802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78797518 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTATGCACCGGGAGACCACCTGG[A/G]GGACATGTGGCCACCATCCCCCACT
Phenotype
MIM: 612543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP36 PubMed Links

Gene Details

Gene
USP36
Gene Name
ubiquitin specific peptidase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321291.1 4019 UTR 3 NP_001308220.1
XM_005257542.2 4019 UTR 3 XP_005257599.1
XM_005257543.2 4019 UTR 3 XP_005257600.1
XM_011525065.1 4019 UTR 3 XP_011523367.1
XM_011525066.2 4019 UTR 3 XP_011523368.1
XM_011525067.2 4019 Intron XP_011523369.1
XM_011525068.1 4019 UTR 3 XP_011523370.1
XM_011525069.2 4019 UTR 3 XP_011523371.1
XM_011525070.1 4019 UTR 3 XP_011523372.1
XM_011525071.2 4019 Intron XP_011523373.1
XM_017024900.1 4019 Intron XP_016880389.1
XM_017024901.1 4019 UTR 3 XP_016880390.1

View Full Product Details