Product Details
- SNP ID
-
rs12410005
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
20
- Location
-
Chr.1:40981264 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCCCATAAGGATGAATGAATGAG[C/T]CTCCCGCCTGATCTTTATGTACACT
- Phenotype
-
MIM: 123860
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTPS1
PubMed Links
Gene Details
- Gene
- CTPS1
- Gene Name
- CTP synthase 1
View Full Product Details