Product Details
- SNP ID
-
rs34998774
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:94909752 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CAAAGACCAATCAGAATGATAACTT[C/T]TTGACTTTTAACTTCCCATTTCAAT
- Phenotype
-
MIM: 602468
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP1R9A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs368119507] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP1R9A
- Gene Name
- protein phosphatase 1 regulatory subunit 9A
View Full Product Details