Product Details
- SNP ID
-
rs2878628
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:52550699 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGACTCTGCACATGTTCTGAGAAGG[A/G]TAACTCGAATTCTGTCAAGTCCTAG
- Phenotype
-
MIM: 606083
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PBRM1
PubMed Links
Gene Details
- Gene
- PBRM1
- Gene Name
- polybromo 1
View Full Product Details