Product Details

SNP ID
rs11132202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183660475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACTGTACTGCGTTAAATTGAA[G/T]ATTAAAATGAATTCCATCCACGTTT
Phenotype
MIM: 614138
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RWDD4 PubMed Links

Gene Details

Gene
RWDD4
Gene Name
RWD domain containing 4
There are no transcripts associated with this gene.

Gene
TRAPPC11
Gene Name
trafficking protein particle complex 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021942.5 Intron NP_068761.4
NM_199053.2 Intron NP_951008.1
XM_017008537.1 Intron XP_016864026.1
XM_017008538.1 Intron XP_016864027.1
XM_017008539.1 Intron XP_016864028.1

View Full Product Details