Product Details
- SNP ID
-
rs766127
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:152992697 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTCCTCGCCTTGCAGTTATCCCAC[A/G]GAGTATCAGGTAAGGATTCTAAGTA
- Phenotype
-
MIM: 606013
MIM: 613542
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73634625] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO5
- Gene Name
- F-box protein 5
There are no transcripts associated with this gene.
- Gene
- MTRF1L
- Gene Name
- mitochondrial translational release factor 1 like
View Full Product Details