Product Details

SNP ID
rs765009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51858581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAAACAAAATGGGGAAGGGGCA[A/G]AAATCTAGAGAAAAAAATAAGCAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100419706 PubMed Links

Gene Details

Gene
LOC100419706
Gene Name
zinc finger protein 84 pseudogene
There are no transcripts associated with this gene.

Gene
ZNF577
Gene Name
zinc finger protein 577
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135590.1 Intron NP_001129062.1
NM_032679.2 Intron NP_116068.2
XM_006723432.3 Intron XP_006723495.1
XM_006723433.2 Intron XP_006723496.1
XM_011527408.2 Intron XP_011525710.1
XM_017027387.1 Intron XP_016882876.1

View Full Product Details