Product Details

SNP ID
hCV348941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13105131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCTGCCCGGAGCTCAGCAGCCC[A/C]TGATGGGATCCTTTCCTGGGATGGG
Phenotype
MIM: 151440 MIM: 164005 MIM: 611669
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LYL1 PubMed Links

Gene Details

Gene
LYL1
Gene Name
LYL1, basic helix-loop-helix family member
There are no transcripts associated with this gene.

Gene
NFIX
Gene Name
nuclear factor I X
There are no transcripts associated with this gene.

Gene
TRMT1
Gene Name
tRNA methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details