Product Details

SNP ID
rs4546432
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:40682448 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CACTTCACTGAGGGAGTGAATGATA[C/T]GCTTAAAATAATCCAGCCGAACCTA
Phenotype
MIM: 601586
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGER4 PubMed Links

Gene Details

Gene
PTGER4
Gene Name
prostaglandin E receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000958.2 Intron NP_000949.1
XM_017009656.1 Intron XP_016865145.1
XM_017009657.1 Intron XP_016865146.1
XM_017009658.1 Intron XP_016865147.1
XM_017009659.1 Intron XP_016865148.1

View Full Product Details