Product Details

SNP ID
rs34376913
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:136484462 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAGCGGCCAGCCATGCCGGGCCCAC[C/T]CACCTGCACACCTGCCTGGCAAGGG
Phenotype
MIM: 190198 MIM: 612854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf163 PubMed Links

Gene Details

Gene
C9orf163
Gene Name
chromosome 9 open reading frame 163
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152571.2 968 Missense Mutation CCC,CTC P5L NP_689784.1
Gene
NOTCH1
Gene Name
notch 1
There are no transcripts associated with this gene.

Gene
SEC16A
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276418.1 968 Intron NP_001263347.1
NM_014866.1 968 Intron NP_055681.1
XM_005266134.2 968 Intron XP_005266191.1
XM_011519246.2 968 Intron XP_011517548.1
XM_011519247.1 968 Intron XP_011517549.1
XM_011519248.2 968 Intron XP_011517550.1
XM_011519249.1 968 Intron XP_011517551.1
XM_011519250.2 968 Intron XP_011517552.1
XM_011519251.1 968 Intron XP_011517553.1
XM_011519252.2 968 Intron XP_011517554.1
XM_011519253.1 968 Intron XP_011517555.1
XM_011519254.1 968 Intron XP_011517556.1
XM_011519255.2 968 Intron XP_011517557.1
XM_011519256.1 968 Intron XP_011517558.1
XM_011519257.1 968 Intron XP_011517559.1
XM_011519258.1 968 Intron XP_011517560.1
XM_011519259.1 968 Intron XP_011517561.1
XM_011519260.2 968 Intron XP_011517562.1
XM_011519261.1 968 Intron XP_011517563.1
XM_011519262.1 968 Intron XP_011517564.1
XM_011519263.2 968 Intron XP_011517565.1
XM_011519264.2 968 Intron XP_011517566.1
XM_017015363.1 968 Intron XP_016870852.1
XM_017015364.1 968 Intron XP_016870853.1

View Full Product Details