Product Details
- SNP ID
-
rs12492588
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:169840923 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATAAGTTAATGATGAAATAAAGACT[A/T]CTGTAAGAGGAGGCAGGAGAGATAA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC31
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79811397] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC31
- Gene Name
- leucine rich repeat containing 31
- Gene
- LRRIQ4
- Gene Name
- leucine rich repeats and IQ motif containing 4
There are no transcripts associated with this gene.
View Full Product Details