Product Details

SNP ID
rs10877722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:61716127 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAAAACAGTATGAGGAACCTAAT[G/T]AGTGCTAAAGAAGTATTAACTATTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM19A2 PubMed Links

Gene Details

Gene
FAM19A2
Gene Name
family with sequence similarity 19 member A2, C-C motif chemokine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178539.4 Intron NP_848634.1
XM_006719370.2 Intron XP_006719433.1
XM_011538272.1 Intron XP_011536574.1
XM_011538274.1 Intron XP_011536576.1

View Full Product Details