Product Details

SNP ID
rs10846103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14977710 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGTTTTAAAATCTATCAATTGTC[C/T]GTTTACACAACTGATAGATTGTGTA
Phenotype
MIM: 601190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE6H PubMed Links

Gene Details

Gene
PDE6H
Gene Name
phosphodiesterase 6H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006205.2 47 Intron NP_006196.1
XM_017019431.1 47 UTR 5 XP_016874920.1

View Full Product Details