Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001521.1 | 148 | Intron | NP_001001521.1 | ||
NM_006759.3 | 148 | Intron | NP_006750.3 | ||
XM_005264537.2 | 148 | UTR 5 | XP_005264594.1 | ||
XM_005264538.1 | 148 | UTR 5 | XP_005264595.1 | ||
XM_006712088.1 | 148 | Intron | XP_006712151.1 | ||
XM_011533091.1 | 148 | Intron | XP_011531393.1 | ||
XM_017004855.1 | 148 | Intron | XP_016860344.1 | ||
XM_017004856.1 | 148 | UTR 5 | XP_016860345.1 | ||
XM_017004857.1 | 148 | Intron | XP_016860346.1 |