Product Details
- SNP ID
-
rs11867841
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:9023715 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGCAAACATTCATTGCCTTCCCCCT[C/G]GTTGGGTTGGACGCTGGGATTCACC
- Phenotype
-
MIM: 601614
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NTN1
PubMed Links
Gene Details
- Gene
- NTN1
- Gene Name
- netrin 1
View Full Product Details