Product Details

SNP ID
rs7252798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21725251 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTCTGTGGCATAATAACACTTCA[C/T]TGAATGCACAATAGTATTTAACATG
Phenotype
MIM: 603982
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF100 PubMed Links

Gene Details

Gene
ZNF100
Gene Name
zinc finger protein 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173531.3 Intron NP_775802.2

View Full Product Details