Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001006935.2 | 59 | Intron | NP_001006936.1 | ||
NM_001006937.2 | 59 | Intron | NP_001006938.1 | ||
NM_001300901.1 | 59 | UTR 5 | NP_001287830.1 | ||
NM_001305840.1 | 59 | Intron | NP_001292769.1 | ||
NM_001305841.1 | 59 | Intron | NP_001292770.1 | ||
NM_001305842.1 | 59 | Intron | NP_001292771.1 | ||
NM_024863.5 | 59 | Intron | NP_079139.4 | ||
XM_017029856.1 | 59 | Intron | XP_016885345.1 | ||
XM_017029857.1 | 59 | Intron | XP_016885346.1 |