Product Details

SNP ID
rs743576
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:11059120 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCCCTCCGCGGGACGCCCCTGGCCT[C/T]GCTAGCACTTTCTCTGACAACCATG
Phenotype
MIM: 605960 MIM: 182891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
There are no transcripts associated with this gene.

Gene
SRM
Gene Name
spermidine synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003132.2 Intron NP_003123.2
XM_017002179.1 Intron XP_016857668.1

View Full Product Details