Product Details
- SNP ID
-
rs471933
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:100992623 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACACATTCACTGTATCCCTCATTGT[A/C]ACCGTTATCCCCCTGCTTCAAAATG
- Phenotype
-
MIM: 615936
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP42
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61995873] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGAP42
- Gene Name
- Rho GTPase activating protein 42
- Gene
- TMEM133
- Gene Name
- transmembrane protein 133
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032021.2 |
544 |
Silent Mutation |
GTA,GTC |
V105V |
NP_114410.1 |
View Full Product Details