Product Details

SNP ID
rs633484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:83262523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTACTTTAGCTTACATCATTT[G/T]GGATAAAAAAAAATTCAGTTTTCCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD42 PubMed Links

Gene Details

Gene
ANKRD42
Gene Name
ankyrin repeat domain 42
There are no transcripts associated with this gene.

Gene
CCDC90B
Gene Name
coiled-coil domain containing 90B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286116.1 Intron NP_001273045.1
NM_001286117.1 Intron NP_001273046.1
NM_001286118.1 Intron NP_001273047.1
NM_001286119.1 Intron NP_001273048.1
NM_001286120.1 Intron NP_001273049.1
NM_021825.4 Intron NP_068597.2
XM_005274154.2 Intron XP_005274211.1
XM_005274156.3 Intron XP_005274213.1
XM_005274159.3 Intron XP_005274216.1
XM_017018103.1 Intron XP_016873592.1
XM_017018104.1 Intron XP_016873593.1

View Full Product Details