Product Details

SNP ID
rs560097
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:65275059 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTATTATATCAGGGTAGTAGAATA[C/A]GGGATTTTTTTTTTAATCTATTTTC
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
POLA2 PubMed Links

Gene Details

Gene
POLA2
Gene Name
DNA polymerase alpha 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002689.3 Intron NP_002680.2
XM_011544877.2 Intron XP_011543179.1
XM_011544878.2 Intron XP_011543180.1
XM_011544879.1 Intron XP_011543181.1
XM_011544881.2 Intron XP_011543183.1
XM_017017452.1 Intron XP_016872941.1

View Full Product Details