Product Details

SNP ID
rs352489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65532281 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAACAAACAAAATCTTTTGTAG[A/G]TGGGGATCAAAGTCCCCGTCTAGAG
Phenotype
MIM: 615940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HACD3 PubMed Links

Gene Details

Gene
HACD3
Gene Name
3-hydroxyacyl-CoA dehydratase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016395.2 Intron NP_057479.2
XM_011521664.1 Intron XP_011519966.1

View Full Product Details