Product Details
- SNP ID
-
rs654527
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:42142307 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGCAGGACACCTGGCTGGCTCC[A/C]TGCGGGCCCTGCTTGGGCCACATCT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PLA2G4F
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111344378] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PLA2G4F
- Gene Name
- phospholipase A2 group IVF
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_213600.3 |
|
Intron |
|
|
NP_998765.3 |
View Full Product Details