Product Details

SNP ID
rs714200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71279675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTATTATTAGTTTATTACTTCA[C/T]CTCCCAGGATATTTTATATGTATAA
Phenotype
MIM: 614349
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF638 PubMed Links

Gene Details

Gene
ZNF638
Gene Name
zinc finger protein 638
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014972.2 Intron NP_001014972.1
NM_001252612.1 Intron NP_001239541.1
NM_001252613.1 Intron NP_001239542.1
NM_014497.4 Intron NP_055312.2
XM_005264263.1 Intron XP_005264320.1
XM_006711989.1 Intron XP_006712052.1
XM_011532768.2 Intron XP_011531070.2
XM_017003809.1 Intron XP_016859298.1
XM_017003810.1 Intron XP_016859299.1
XM_017003811.1 Intron XP_016859300.1
XM_017003812.1 Intron XP_016859301.1

View Full Product Details