Product Details

SNP ID
rs755770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130166749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACCACGAGACCCAGAGTTCTGAG[C/T]TGACAGTGAGCCCAGGAGAGATCAG
Phenotype
MIM: 606797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST14 PubMed Links

Gene Details

Gene
ST14
Gene Name
suppression of tumorigenicity 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021978.3 Intron NP_068813.1

View Full Product Details