Product Details

SNP ID
rs753991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98123010 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTTCTGTAGCAAACCCAGATCAC[C/G]CCACAGGTAGTTTTCTCTTAGGTCA
Phenotype
MIM: 602159 MIM: 606556
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CORO2A PubMed Links

Gene Details

Gene
CORO2A
Gene Name
coronin 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003389.3 3608 UTR 3 NP_003380.3
NM_052820.3 3608 UTR 3 NP_438171.1
XM_011518986.2 3608 UTR 3 XP_011517288.1
Gene
TRIM14
Gene Name
tripartite motif containing 14
There are no transcripts associated with this gene.

View Full Product Details