Product Details
- SNP ID
-
rs510160
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:113168167 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTTTGTTTAAACAGTATCCTGT[A/G]AAGAAGTTTAAAGAATAAGGAAAAA
- Phenotype
-
MIM: 613713
MIM: 176895
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PCID2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2146750] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCID2
- Gene Name
- PCI domain containing 2
- Gene
- PROZ
- Gene Name
- protein Z, vitamin K dependent plasma glycoprotein
View Full Product Details