Product Details

SNP ID
rs691391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39084082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCGCCTGAGGGACGAGCGCAGG[C/G]TTACCTGCCTGGGCTCTCACCTGGT
Phenotype
MIM: 610490
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACP7 PubMed Links

Gene Details

Gene
ACP7
Gene Name
acid phosphatase 7, tartrate resistant (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004318.2 Intron NP_001004318.2
XM_011526966.2 Intron XP_011525268.1
XM_011526967.2 Intron XP_011525269.1
XM_011526969.1 Intron XP_011525271.1
XM_017026807.1 Intron XP_016882296.1
XM_017026808.1 Intron XP_016882297.1

View Full Product Details