Product Details

SNP ID
rs595954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36497057 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGAAAGATAAGAATAACTAAAAC[C/G]TGAAGTTAGTGCAACTCCCCCCATG
Phenotype
MIM: 615932 MIM: 606964
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCTD20 PubMed Links

Gene Details

Gene
KCTD20
Gene Name
potassium channel tetramerization domain containing 20
There are no transcripts associated with this gene.

Gene
STK38
Gene Name
serine/threonine kinase 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305102.1 Intron NP_001292031.1
NM_007271.3 Intron NP_009202.1
XM_006714988.3 Intron XP_006715051.1
XM_006714989.3 Intron XP_006715052.1
XM_017010226.1 Intron XP_016865715.1

View Full Product Details