Product Details
- SNP ID
-
rs745879
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
30
- Location
-
Chr.1:41479907 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCTGCTACAATTCTACTCATTCTTC[A/G]AGGTCCAGTTCAAGGTAGTCCAGCC
- Phenotype
-
MIM: 131241
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EDN2
PubMed Links
Gene Details
- Gene
- EDN2
- Gene Name
- endothelin 2
View Full Product Details